Gene Transcripts

Gene Symbol
Ctrc
Entrez Gene ID
76701
Gene Name
chymotrypsin C (caldecrin)
Gene Aliases
1810044E12Rik
Chromosome Location
Chr. 4: 141838239 - 141846359 on Build GRCm38
UniGene ID
Mm.489680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033875.2 NP_001029047.1 8 946
GenBank mRNA BC103715.1 - 8 944

View Full Product Details