Gene Transcripts

Gene Symbol
Olfr59
Entrez Gene ID
18359
Gene Name
olfactory receptor 59
Gene Aliases
IH3, MOR133-3P
Chromosome Location
Chr. 11: 74283752 - 74289646 on Build GRCm38
UniGene ID
Mm.300072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011002.2 NP_035132.2 2 792
GenBank mRNA AF102538.1 - 1 274
BC139159.1 - 1 552 8862181
BC139161.1 - 1 552 8862183

View Full Product Details