Gene Transcripts

Gene Symbol
Hs3st3b1
Entrez Gene ID
54710
Gene Name
heparan sulfate (glucosamine) 3-O-sulfotransferase 3B1
Gene Aliases
3-OST-3B, 3OST3B1, 3Ost3b, AU041882, AW536289, Hs3st3b, m3-OST-3B
Chromosome Location
Chr. 11: 63884693 - 63922284 on Build GRCm38
UniGene ID
Mm.274426
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018805.2 NP_061275.2 2 1592
GenBank mRNA AF168992.1 - 2 1550
AK085242.1 - 2 1601
AK171996.1 - 2 1586
AK172015.1 - 2 1591
AK172271.1 - 2 1593

View Full Product Details