Gene Transcripts

Gene Symbol
VAT1L
Entrez Gene ID
57687
Gene Name
vesicle amine transport 1-like
Gene Aliases
Chromosome Location
Chr. 16: 77788564 - 77980107 on Build GRCh38
UniGene ID
Hs.461405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020927.2 NP_065978.1 2 395
GenBank mRNA AB046796.1 - 2 365
AK290153.1 - 2 373
BC033537.1 - 2 373 4821265
HY132852.1 - 2 396

View Full Product Details