Gene Transcripts

Gene Symbol
Oxct2b
Entrez Gene ID
353371
Gene Name
3-oxoacid CoA transferase 2B
Gene Aliases
Scot-t2
Chromosome Location
Chr. 4: 123116248 - 123118000 on Build GRCm38
UniGene ID
Mm.189660
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181859.3 NP_862907.2 1 1631
GenBank mRNA AB049996.1 - 1 1621
BC113758.1 - 1 1614 40047052

View Full Product Details