Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320755.1 | NP_001307684.1 | 4 | 508 | |
NM_003170.4 | NP_003161.2 | 3 | 406 | ||
XM_017024978.1 | XP_016880467.1 | 5 | 567 | ||
XM_017024979.1 | XP_016880468.1 | 3 | 644 | ||
XM_017024980.1 | XP_016880469.1 | 2 | 338 | ||
GenBank mRNA | AL833607.1 | - | 4 | 509 | |
BC136522.1 | - | 3 | 318 | 9020512 | |
BC136524.1 | - | 3 | 318 | 9020514 | |
BC150268.1 | - | 3 | 320 | 9007230 | |
D79984.1 | - | 3 | 320 |