Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021626.2 | NP_067639.1 | 2 | 202 | |
XM_005257557.2 | XP_005257614.1 | 2 | 184 | ||
GenBank mRNA | AF113214.1 | - | 2 | 127 | |
AF282618.1 | - | 2 | 181 | ||
AK027373.1 | - | 2 | 175 | ||
AK298161.1 | - | 2 | 121 | ||
AY358559.1 | - | 2 | 182 | ||
BC072405.1 | - | 2 | 152 | 6137668 | |
DC344612.1 | - | 2 | 202 |