Gene Transcripts
- Gene Symbol
- THADA
- Entrez Gene ID
- 63892
- Gene Name
- THADA, armadillo repeat containing
- Gene Aliases
- ARMC13, GITA
- Chromosome Location
- Chr. 2: 43230836 - 43596046 on Build GRCh38
- UniGene ID
- Hs.369592
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001083953.1 |
NP_001077422.1
|
31, 32 |
4785 |
|
NM_022065.4 |
NP_071348.3
|
31, 32 |
4573 |
|
NR_073394.1 |
|
29, 30 |
4383 |
|
XM_006712061.3 |
XP_006712124.1
|
32, 33 |
4620 |
|
XM_006712062.1 |
XP_006712125.1
|
31, 32 |
4570 |
|
XM_006712063.1 |
XP_006712126.1
|
31, 32 |
4453 |
|
XM_006712064.2 |
XP_006712127.1
|
31, 32 |
4573 |
|
XM_006712065.1 |
XP_006712128.1
|
29, 30 |
4351 |
|
XM_006712066.1 |
XP_006712129.1
|
29, 30 |
4210 |
|
XM_006712067.1 |
XP_006712130.1
|
29, 30 |
4207 |
|
XM_006712069.3 |
XP_006712132.1
|
16, 17 |
2081 |
|
XM_017004674.1 |
XP_016860163.1
|
31, 32 |
4450 |
|
XM_017004675.1 |
XP_016860164.1
|
30, 31 |
4325 |
|
GenBank mRNA |
AB051554.2 |
- |
31, 32 |
4785 |
|
AF323176.1 |
- |
4, 5 |
503 |
|
AK025445.1 |
- |
16, 17 |
2152 |
|
AK025530.1 |
- |
6, 7 |
646 |
|
AK290023.1 |
- |
31, 32 |
4573 |
|
AY149629.1 |
- |
31, 32 |
4565 |
|
AY149633.1 |
- |
29, 30 |
4375 |
|
BC007743.1 |
- |
2, 3 |
167 |
|
BC025773.1 |
- |
7, 8 |
786 |
|
BC105976.1 |
- |
10, 11 |
1083 |
|
BX648155.1 |
- |
31, 32 |
4569 |
|
View Full Product Details