Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001168474.1 | NP_001161946.1 | 6 | 564 | |
NM_024885.3 | NP_079161.3 | 6 | 732 | ||
XM_005262152.1 | XP_005262209.1 | 6 | 564 | ||
XM_006724664.1 | XP_006724727.1 | 6 | 731 | ||
GenBank mRNA | AF285595.1 | - | 6 | 566 | |
AK026810.1 | - | 5 | 710 | ||
AK308261.1 | - | 6 | 734 | ||
BC043391.1 | - | 6 | 718 | 6154641 | |
DB042169.1 | - | 6 | 564 |