Gene Transcripts
- Gene Symbol
- RUFY1
- Entrez Gene ID
- 80230
- Gene Name
- RUN and FYVE domain containing 1
- Gene Aliases
- RABIP4, ZFYVE12
- Chromosome Location
- Chr. 5: 179550551 - 179610026 on Build GRCh38
- UniGene ID
- Hs.306769
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040451.2 |
NP_001035541.1
|
5, 6 |
919 |
|
NM_001040452.2 |
NP_001035542.1
|
6, 7 |
730 |
|
NM_025158.4 |
NP_079434.3
|
6, 7 |
905 |
|
XM_005265993.4 |
XP_005266050.1
|
3, 4 |
276 |
|
XM_006714922.3 |
XP_006714985.1
|
2, 3 |
476 |
|
XM_017009892.1 |
XP_016865381.1
|
4, 5 |
702 |
|
XM_017009894.1 |
XP_016865383.1
|
6, 7 |
638 |
|
XM_017009895.1 |
XP_016865384.1
|
5, 6 |
710 |
|
GenBank mRNA |
AB209507.1 |
- |
6, 7 |
630 |
|
AF312367.1 |
- |
6, 7 |
896 |
|
AF361055.2 |
- |
6, 7 |
878 |
|
AK075021.1 |
- |
5, 6 |
818 |
|
AK291926.1 |
- |
5, 6 |
919 |
|
BC032571.1 |
- |
6, 7 |
649 |
5243223 |
View Full Product Details