Gene Transcripts
- Gene Symbol
- TSPAN14
- Entrez Gene ID
- 81619
- Gene Name
- tetraspanin 14
- Gene Aliases
- DC-TM4F2, TM4SF14
- Chromosome Location
- Chr. 10: 80454271 - 80522635 on Build GRCh38
- UniGene ID
- Hs.718943
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001128309.1 |
NP_001121781.1
|
4, 5 |
360 |
|
NM_030927.2 |
NP_112189.2
|
7, 8 |
729 |
|
XM_006717992.3 |
XP_006718055.1
|
7, 8 |
885 |
|
XM_011540219.2 |
XP_011538521.1
|
9, 10 |
919 |
|
XM_011540220.2 |
XP_011538522.1
|
9, 10 |
841 |
|
XM_011540221.2 |
XP_011538523.1
|
8, 9 |
703 |
|
XM_011540222.2 |
XP_011538524.1
|
8, 9 |
881 |
|
XM_017016707.1 |
XP_016872196.1
|
9, 10 |
877 |
|
XM_017016708.1 |
XP_016872197.1
|
8, 9 |
838 |
|
XM_017016709.1 |
XP_016872198.1
|
8, 9 |
886 |
|
XM_017016710.1 |
XP_016872199.1
|
8, 9 |
837 |
|
XM_017016711.1 |
XP_016872200.1
|
8, 9 |
702 |
|
XM_017016712.1 |
XP_016872201.1
|
8, 9 |
905 |
|
XM_017016713.1 |
XP_016872202.1
|
7, 8 |
725 |
|
GenBank mRNA |
AF311903.1 |
- |
7, 8 |
694 |
|
AK295321.1 |
- |
4, 5 |
331 |
|
AL136638.1 |
- |
7, 8 |
700 |
|
BC002920.2 |
- |
6, 7 |
624 |
3954042 |
BC093016.1 |
- |
7, 8 |
729 |
30528940 |
BI918996.1 |
- |
4, 5 |
383 |
|
DC300676.1 |
- |
4, 5 |
332 |
|
View Full Product Details