Gene Transcripts
- Gene Symbol
- MRO
- Entrez Gene ID
- 83876
- Gene Name
- maestro
- Gene Aliases
- B29, C18orf3
- Chromosome Location
- Chr. 18: 50795120 - 50825413 on Build GRCh38
- UniGene ID
- Hs.30495
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001127174.1 |
NP_001120646.1
|
2, 3 |
187 |
|
NM_031939.3 |
NP_114145.2
|
3, 4 |
401 |
|
XM_005258365.3 |
XP_005258422.1
|
2, 3 |
211 |
|
XM_011526226.2 |
XP_011524528.1
|
3, 4 |
338 |
|
XM_017026035.1 |
XP_016881524.1
|
2, 3 |
178 |
|
XM_017026036.1 |
XP_016881525.1
|
3, 4 |
338 |
|
XM_017026037.1 |
XP_016881526.1
|
2, 3 |
327 |
|
GenBank mRNA |
AB042647.1 |
- |
3, 4 |
265 |
|
AK054702.1 |
- |
3, 4 |
335 |
|
DC403805.1 |
- |
2, 3 |
183 |
|
View Full Product Details