Gene Transcripts

Gene Symbol
PRAC1
Entrez Gene ID
84366
Gene Name
prostate cancer susceptibility candidate 1
Gene Aliases
C17orf92, PRAC
Chromosome Location
Chr. 17: 48721719 - 48722520 on Build GRCh38
UniGene ID
Hs.116467
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032391.2 NP_115767.1 2 297
GenBank mRNA AF331165.1 - 2 297
BC030950.1 - 2 295 4739311
BU535394.1 - 2 300

View Full Product Details