Gene Transcripts

Gene Symbol
MAFG
Entrez Gene ID
4097
Gene Name
MAF bZIP transcription factor G
Gene Aliases
hMAF
Chromosome Location
Chr. 17: 81918269 - 81927711 on Build GRCh38
UniGene ID
Hs.252229
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002359.3 NP_002350.1 3 4153
NM_032711.3 NP_116100.2 3 4185
XM_011523578.1 XP_011521880.1 3 5088
GenBank mRNA AK130699.1 - 1 1126
BC006143.1 - 1 536
BC103722.1 - 1 811

View Full Product Details