Gene Transcripts
- Gene Symbol
- UTP4
- Entrez Gene ID
- 84916
- Gene Name
- UTP4, small subunit processome component
- Gene Aliases
- CIRH1A, CIRHIN, NAIC, TEX292
- Chromosome Location
- Chr. 16: 69132596 - 69169034 on Build GRCh38
- UniGene ID
- Hs.461113
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318391.1 |
NP_001305320.1
|
2, 3 |
203 |
|
NM_032830.2 |
NP_116219.2
|
2, 3 |
257 |
|
GenBank mRNA |
AB075868.2 |
- |
2, 3 |
195 |
|
AK027634.1 |
- |
2, 3 |
209 |
|
AK027675.1 |
- |
2, 3 |
206 |
|
AK074795.1 |
- |
2, 3 |
223 |
|
AK310104.1 |
- |
2, 3 |
150 |
|
BC009348.2 |
- |
2, 3 |
189 |
4131625 |
BX647265.1 |
- |
2, 3 |
171 |
|
DA277545.1 |
- |
2, 3 |
203 |
|
DC368305.1 |
- |
2, 3 |
257 |
|
View Full Product Details