Gene Transcripts
- Gene Symbol
- TRIM55
- Entrez Gene ID
- 84675
- Gene Name
- tripartite motif containing 55
- Gene Aliases
- MURF-2, RNF29, muRF2
- Chromosome Location
- Chr. 8: 66112667 - 66175485 on Build GRCh38
- UniGene ID
- Hs.85524
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_033058.2 |
NP_149047.2
|
1, 2 |
402 |
|
NM_184085.1 |
NP_908973.1
|
1, 2 |
402 |
|
NM_184086.1 |
NP_908974.1
|
1, 2 |
402 |
|
NM_184087.1 |
NP_908975.1
|
1, 2 |
402 |
|
XM_005251316.4 |
XP_005251373.1
|
3, 4 |
1427 |
|
XM_011517613.2 |
XP_011515915.1
|
3, 4 |
1427 |
|
XM_011517614.2 |
XP_011515916.1
|
3, 4 |
1350 |
|
XM_011517615.2 |
XP_011515917.1
|
3, 4 |
1427 |
|
XM_011517616.2 |
XP_011515918.1
|
3, 4 |
1427 |
|
XM_017013908.1 |
XP_016869397.1
|
2 |
120 |
|
GenBank mRNA |
AJ243488.1 |
- |
1, 2 |
402 |
|
AJ243489.1 |
- |
1, 2 |
402 |
|
AJ277493.1 |
- |
1, 2 |
402 |
|
AJ291712.1 |
- |
1, 2 |
308 |
|
AJ431704.1 |
- |
1, 2 |
402 |
|
AK091310.1 |
- |
1, 2 |
552 |
|
AK091728.1 |
- |
1, 2 |
222 |
|
BC007750.2 |
- |
1, 2 |
308 |
4110783 |
BT007212.1 |
- |
1, 2 |
128 |
|
View Full Product Details