Gene Transcripts

Gene Symbol
PIK3IP1
Entrez Gene ID
113791
Gene Name
phosphoinositide-3-kinase interacting protein 1
Gene Aliases
HGFL, hHGFL(S)
Chromosome Location
Chr. 22: 31281593 - 31292534 on Build GRCh38
UniGene ID
Hs.26670
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135911.1 NP_001129383.1 5 1338
NM_052880.4 NP_443112.2 6 1417
GenBank mRNA AK074688.1 - 6 1363
AK093768.1 - 4 1926
AK226089.1 - 6 1297
AK300990.1 - 7 1275
BC011049.2 - 6 1362 4180134
BC041903.1 - 4 1714

View Full Product Details