Gene Transcripts
- Gene Symbol
- SPSB3
- Entrez Gene ID
- 90864
- Gene Name
- splA/ryanodine receptor domain and SOCS box containing 3
- Gene Aliases
- C16orf31, SSB3
- Chromosome Location
- Chr. 16: 1776712 - 1782605 on Build GRCh38
- UniGene ID
- Hs.592080
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001324081.1 |
NP_001311010.1
|
3, 4 |
371 |
|
NM_080861.3 |
NP_543137.2
|
3, 4 |
393 |
|
XM_005255673.4 |
XP_005255730.1
|
3, 4 |
583 |
|
GenBank mRNA |
AF403028.1 |
- |
2, 3 |
302 |
|
AK074190.1 |
- |
3, 4 |
357 |
|
BC007588.1 |
- |
3, 4 |
339 |
3342825 |
BC024244.1 |
- |
3, 4 |
357 |
4798873 |
BC035065.1 |
- |
3, 4 |
350 |
|
BC047441.1 |
- |
3, 4 |
335 |
5303706 |
BI768095.1 |
- |
3, 4 |
347 |
|
BM836743.1 |
- |
3, 4 |
418 |
|
View Full Product Details