Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001256604.1 | NP_001243533.1 | 6 | 1147 | |
NM_001256605.1 | NP_001243534.1 | 5 | 1036 | ||
NM_001256608.1 | NP_001243537.1 | 5 | 1037 | ||
NM_001256609.1 | NP_001243538.1 | 6 | 1523 | ||
NM_144580.2 | NP_653181.1 | 6 | 1294 | ||
XM_011509117.2 | XP_011507419.1 | 5 | 953 | ||
XM_017000149.1 | XP_016855638.1 | 6 | 1154 | ||
GenBank mRNA | AK075349.1 | - | 6 | 1263 | |
AK296157.1 | - | 6 | 1491 | ||
AY358450.1 | - | 6 | 1267 | ||
BC011575.1 | - | 6 | 1277 | ||
BC018757.1 | - | 6 | 1263 | 4850051 | |
BC036340.1 | - | 6 | 1210 |