Gene Transcripts

Gene Symbol
TMEM263
Entrez Gene ID
90488
Gene Name
transmembrane protein 263
Gene Aliases
C12orf23
Chromosome Location
Chr. 12: 106955535 - 106974037 on Build GRCh38
UniGene ID
Hs.257664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319661.1 NP_001306590.1 2, 3 422
NM_001319662.1 NP_001306591.1 2, 3 443
NM_001319663.1 NP_001306592.1 4, 5 588
NM_001319664.1 NP_001306593.1 4, 5 606
NM_152261.3 NP_689474.1 3, 4 490
XM_017020198.1 XP_016875687.1 3, 4 677
GenBank mRNA AK001731.1 - 3, 4 305
AK021783.1 - 1, 2 105
AK022770.1 - 2, 3 204
AK024021.1 - 3, 4 317
BC020522.1 - 3, 4 285 3916886
CX166555.1 - 4, 5 482
DA609590.1 - 2, 3 443
DB091654.1 - 3, 4 490
DB233011.1 - 4, 5 411

View Full Product Details