Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193646.1 | NP_001180575.1 | 3 | 1746 | |
NM_001290746.1 | NP_001277675.1 | 3 | 1717 | ||
NM_012068.5 | NP_036200.2 | 4 | 1951 | ||
XM_011526629.2 | XP_011524931.1 | 3 | 1653 | ||
GenBank mRNA | AB021663.2 | - | 3 | 1688 | |
AB073613.1 | - | 3 | 1684 | ||
AK091985.1 | - | 2 | 1660 |