Gene Transcripts

Gene Symbol
LPAR1
Entrez Gene ID
1902
Gene Name
lysophosphatidic acid receptor 1
Gene Aliases
EDG2, GPR26, Gpcr26, LPA1, Mrec1.3, VZG1, edg-2, rec.1.3, vzg-1
Chromosome Location
Chr. 9: 110873252 - 111039259 on Build GRCh38
UniGene ID
Hs.126667
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001401.3 NP_001392.2 3 793
NM_057159.2 NP_476500.1 4 871
XM_005251781.3 XP_005251838.1 4 1016
XM_005251782.3 XP_005251839.1 4 1110
XM_017014383.1 XP_016869872.1 4 910
XM_017014384.1 XP_016869873.1 5 1284
XM_017014385.1 XP_016869874.1 6 877
XM_017014386.1 XP_016869875.1 6 1177
XM_017014387.1 XP_016869876.1 6 913
XM_017014388.1 XP_016869877.1 6 1096
XM_017014389.1 XP_016869878.1 5 839
XM_017014390.1 XP_016869879.1 6 874
XM_017014391.1 XP_016869880.1 6 898
XM_017014392.1 XP_016869881.1 5 1141
XM_017014393.1 XP_016869882.1 6 937
XM_017014394.1 XP_016869883.1 5 1066
XM_017014395.1 XP_016869884.1 5 894
XM_017014396.1 XP_016869885.1 4 836
XM_017014397.1 XP_016869886.1 5 767
XM_017014398.1 XP_016869887.1 5 1010
XM_017014399.1 XP_016869888.1 5 765
XM_017014400.1 XP_016869889.1 6 1126
XM_017014401.1 XP_016869890.1 5 797
XM_017014402.1 XP_016869891.1 5 854
XM_017014403.1 XP_016869892.1 6 852
XM_017014404.1 XP_016869893.1 4 1050
XM_017014405.1 XP_016869894.1 6 1039
XM_017014406.1 XP_016869895.1 5 773
XM_017014407.1 XP_016869896.1 5 829
GenBank mRNA AK022808.1 - 3 793
AK290287.1 - 5 1146
AK296374.1 - 2 474
AK298590.1 - 2 450
AK299279.1 - 2 524
AY322546.1 - 2 445
BC030615.2 - 4 700 4815446
BC036034.1 - 4 830 5272431
BC073167.1 - 5 740 6193969
U78192.1 - 3 567
U80811.1 - 2 471
Y09479.1 - 3 567

View Full Product Details