Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153486.3 | NP_705690.2 | 3 | 325 | |
NM_194436.2 | NP_919417.1 | 3 | 325 | ||
GenBank mRNA | AY092767.1 | - | 3 | 330 | |
BC040279.1 | - | 3 | 290 | 5162826 | |
BC047902.1 | - | 3 | 320 | 5762691 | |
BM544922.1 | - | 3 | 331 |