Gene Transcripts

Gene Symbol
C17orf74
Entrez Gene ID
201243
Gene Name
chromosome 17 open reading frame 74
Gene Aliases
Chromosome Location
Chr. 17: 7424874 - 7427568 on Build GRCh38
UniGene ID
Hs.380704
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175734.4 NP_783861.3 3 1569
XM_006721471.3 XP_006721534.1 3 1304
XM_011523722.2 XP_011522024.1 3 2157
GenBank mRNA BC031286.1 - 3 1569
BC044816.1 - 3 1206
BC048010.1 - 3 1569

View Full Product Details