Gene Transcripts

Gene Symbol
NLRP9
Entrez Gene ID
338321
Gene Name
NLR family pyrin domain containing 9
Gene Aliases
CLR19.1, NALP9, NOD6, PAN12
Chromosome Location
Chr. 19: 55708432 - 55738402 on Build GRCh38
UniGene ID
Hs.661568
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176820.3 NP_789790.2 1 234
GenBank mRNA AY154464.1 - 1 206
BC136583.1 - 1 226 9020573

View Full Product Details