Gene Transcripts

Gene Symbol
TRAM1L1
Entrez Gene ID
133022
Gene Name
translocation associated membrane protein 1-like 1
Gene Aliases
Chromosome Location
Chr. 4: 117083554 - 117085580 on Build GRCh38
UniGene ID
Hs.570737
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152402.2 NP_689615.2 1 940
GenBank mRNA AK074617.1 - 1 936
BC030831.1 - 1 936 4796626
BX423039.2 - 1 873

View Full Product Details