Gene Transcripts
- Gene Symbol
- NFKBIB
- Entrez Gene ID
- 4793
- Gene Name
- NFKB inhibitor beta
- Gene Aliases
- IKBB, TRIP9
- Chromosome Location
- Chr. 19: 38899503 - 38908894 on Build GRCh38
- UniGene ID
- Hs.9731
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001243116.1 |
NP_001230045.1
|
2, 3 |
139 |
|
NM_002503.4 |
NP_002494.2
|
2, 3 |
381 |
|
NR_040515.1 |
|
2, 3 |
325 |
|
XM_006723226.2 |
XP_006723289.2
|
2, 3 |
317 |
|
XM_006723227.3 |
XP_006723290.3
|
2, 3 |
280 |
|
GenBank mRNA |
AB209784.1 |
- |
2, 3 |
347 |
|
AK057862.1 |
- |
2, 3 |
276 |
|
AK290569.1 |
- |
2, 3 |
332 |
|
BC015528.1 |
- |
2, 3 |
312 |
3903794 |
BI520169.1 |
- |
2, 3 |
139 |
|
BM008311.1 |
- |
2, 3 |
333 |
|
BM449613.1 |
- |
2, 3 |
336 |
|
BQ440907.1 |
- |
2, 3 |
113 |
|
BT006743.1 |
- |
2, 3 |
278 |
|
DA633677.1 |
- |
2, 3 |
403 |
|
View Full Product Details