Gene Transcripts

Gene Symbol
Lgals5
Entrez Gene ID
25475
Gene Name
lectin, galactose binding, soluble 5
Gene Aliases
Chromosome Location
Chr. 10: 66226015 - 66229293 on Build Rnor_6.0
UniGene ID
Rn.64588
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012976.2 NP_037108.1 2 129
GenBank mRNA FQ225275.1 - 2 123
FQ225984.1 - 2 145
FQ226981.1 - 2 150
FQ227276.1 - 2 144
FQ228081.1 - 2 149
FQ230182.1 - 2 149
FQ231538.1 - 2 148
FQ231564.1 - 2 159
FQ232247.1 - 2 170
FQ232513.1 - 2 152
FQ233614.1 - 2 170
FQ234601.1 - 2 152
FQ234845.1 - 2 172
L21711.1 - 2 138
L36862.1 - 2 129

View Full Product Details