Gene Transcripts

Gene Symbol
Cdipt
Entrez Gene ID
192260
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Gene Aliases
Pis, Pis1
Chromosome Location
Chr. 1: 198409186 - 198413497 on Build Rnor_6.0
UniGene ID
Rn.10598
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138899.2 NP_620254.1 6 1586
XM_006230208.1 XP_006230270.1 7 1622
GenBank mRNA BC070876.1 - 6 1586
D82928.1 - 6 1334

View Full Product Details