Gene Transcripts

Gene Symbol
Nfyc
Entrez Gene ID
25337
Gene Name
nuclear transcription factor-Y gamma
Gene Aliases
CBF-C
Chromosome Location
Chr. 5: 139687442 - 139759741 on Build Rnor_6.0
UniGene ID
Rn.1457
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012866.2 NP_036998.1 4, 5 497
XM_006238785.2 XP_006238847.1 4, 5 470
GenBank mRNA BC093619.1 - 4, 5 497
U17607.1 - 4, 5 385

View Full Product Details