Gene Transcripts

Gene Symbol
Camk2a
Entrez Gene ID
25400
Gene Name
calcium/calmodulin-dependent protein kinase II alpha
Gene Aliases
PK2CDD, PKCCD
Chromosome Location
Chr. 18: 56193978 - 56295869 on Build Rnor_6.0
UniGene ID
Rn.107499
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012920.1 NP_037052.1 10 784
GenBank mRNA J02942.1 - 10 784
M16960.1 - 4 350

View Full Product Details