Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001303433.1 | NP_001290362.1 | 7 | 627 | |
NM_003902.4 | NP_003893.2 | 6 | 564 | ||
NR_130152.1 | 6 | 561 | |||
XM_011542391.2 | XP_011540693.1 | 7 | 498 | ||
XM_011542392.2 | XP_011540694.1 | 7 | 529 | ||
XM_011542393.2 | XP_011540695.1 | 6 | 469 | ||
XM_017002739.1 | XP_016858228.1 | 8 | 520 | ||
XM_017002740.1 | XP_016858229.1 | 7 | 498 | ||
XM_017002741.1 | XP_016858230.1 | 7 | 513 | ||
XM_017002742.1 | XP_016858231.1 | 7 | 529 | ||
XM_017002743.1 | XP_016858232.1 | 6 | 470 | ||
XM_017002744.1 | XP_016858233.1 | 6 | 470 | ||
GenBank mRNA | AK300032.1 | - | 7 | 495 | |
AK301581.1 | - | 6 | 429 | ||
AK303568.1 | - | 6 | 428 | ||
BC010083.2 | - | 6 | 399 | ||
BC017247.2 | - | 6 | 425 | 4891583 | |
BQ649331.1 | - | 6 | 452 | ||
U05040.2 | - | 6 | 389 |