Gene Transcripts

Gene Symbol
Kcnj13
Entrez Gene ID
100040591
Gene Name
potassium inwardly-rectifying channel, subfamily J, member 13
Gene Aliases
Chromosome Location
Chr. 1: 87386284 - 87394729 on Build GRCm38
UniGene ID
Mm.443539
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110227.1 NP_001103697.1 3 1021
GenBank mRNA AK210794.1 - 1 114

View Full Product Details