Gene Transcripts

Gene Symbol
Mki67
Entrez Gene ID
17345
Gene Name
antigen identified by monoclonal antibody Ki 67
Gene Aliases
D630048A14Rik, Ki-67, Ki67
Chromosome Location
Chr. 7: 135689782 - 135716408 on Build GRCm38
UniGene ID
Mm.4078
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081117.2 NP_001074586.2 12 3377
GenBank mRNA AK082099.1 - 12 3296
X82786.1 - 12 3485

View Full Product Details