Gene Transcripts
- Gene Symbol
- Ldhc
- Entrez Gene ID
- 29634
- Gene Name
- lactate dehydrogenase C
- Gene Aliases
- Ldh3
- Chromosome Location
- Chr. 1: 102915041 - 102931843 on Build Rnor_6.0
- UniGene ID
- Rn.9984
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017266.2 |
NP_058962.2
|
2, 3 |
237 |
|
XM_006229233.2 |
XP_006229295.1
|
2, 3 |
308 |
|
XM_008759344.1 |
XP_008757566.1
|
2, 3 |
184 |
|
XM_008759345.1 |
XP_008757567.1
|
2, 3 |
184 |
|
GenBank mRNA |
BC078862.1 |
- |
2, 3 |
237 |
|
JQ173138.1 |
- |
1, 2 |
134 |
|
JQ173140.1 |
- |
1, 2 |
134 |
|
JQ173141.1 |
- |
1, 2 |
134 |
|
JQ409364.1 |
- |
1, 2 |
134 |
|
JQ409366.1 |
- |
1, 2 |
134 |
|
JQ409367.1 |
- |
1, 2 |
134 |
|
JQ409368.1 |
- |
1, 2 |
134 |
|
JQ409369.1 |
- |
1, 2 |
134 |
|
U07177.1 |
- |
1, 2 |
138 |
|
View Full Product Details