Gene Transcripts

Gene Symbol
Mcu
Entrez Gene ID
215999
Gene Name
mitochondrial calcium uniporter
Gene Aliases
2010012O16Rik, AV064928, C10orf42, Ccdc109a, D130073L02Rik, Gm64
Chromosome Location
Chr. 10: 59446984 - 59616692 on Build GRCm38
UniGene ID
Mm.296470
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033259.4 NP_001028431.2 8 1653
GenBank mRNA AK008216.1 - 1 39

View Full Product Details