Gene Transcripts

Gene Symbol
Taf4a
Entrez Gene ID
228980
Gene Name
TAF4A RNA polymerase II, TATA box binding protein (TBP)-associated factor
Gene Aliases
135kDa, AI450312, TAFII130, TAFII135, Taf2c1
Chromosome Location
Chr. 2: 179912146 - 179976646 on Build GRCm38
UniGene ID
Mm.235542
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081092.1 NP_001074561.1 2 1273
GenBank mRNA AY038601.1 - 2 129

View Full Product Details