Gene Transcripts

Gene Symbol
Cog5
Entrez Gene ID
238123
Gene Name
component of oligomeric golgi complex 5
Gene Aliases
5430405C01Rik, C87247, D18362, GOLTC1, GTC90
Chromosome Location
Chr. 12: 31654868 - 31937630 on Build GRCm38
UniGene ID
Mm.173068
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163126.1 NP_001156598.1 9 928
GenBank mRNA AK030656.1 - 9 929
CA316530.1 - 6 583

View Full Product Details