Gene Transcripts

Gene Symbol
Lonrf1
Entrez Gene ID
244421
Gene Name
LON peptidase N-terminal domain and ring finger 1
Gene Aliases
Chromosome Location
Chr. 8: 36216064 - 36249516 on Build GRCm38
UniGene ID
Mm.324032
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081150.1 NP_001074619.1 12 3690
GenBank mRNA AK138879.1 - 1 620
BC094431.1 - 9 2496

View Full Product Details