Gene Transcripts

Gene Symbol
Gsk3a
Entrez Gene ID
50686
Gene Name
glycogen synthase kinase 3 alpha
Gene Aliases
Chromosome Location
Chr. 1: 82097244 - 82108203 on Build Rnor_6.0
UniGene ID
Rn.36807
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017344.1 NP_059040.1 2, 3 590
XM_008758972.1 XP_008757194.1 2, 3 578
GenBank mRNA X53427.1 - 2, 3 590

View Full Product Details