Gene Transcripts

Gene Symbol
Siglec15
Entrez Gene ID
620235
Gene Name
sialic acid binding Ig-like lectin 15
Gene Aliases
Cd33l3, EG620235, SIGLEC-I
Chromosome Location
Chr. 18: 78042499 - 78057435 on Build GRCm38
UniGene ID
Mm.35607
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001101038.1 NP_001094508.1 4 513
GenBank mRNA AY971517.1 - 4 513

View Full Product Details