Gene Transcripts
- Gene Symbol
- Chrm5
- Entrez Gene ID
- 53949
- Gene Name
- cholinergic receptor, muscarinic 5
- Gene Aliases
- Chromosome Location
- Chr. 3: 103966451 - 104018815 on Build Rnor_6.0
- UniGene ID
- Rn.44293
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017362.4 |
NP_059058.1
|
|
2175 |
|
GenBank mRNA |
M22926.1 |
- |
|
2175 |
|
View Full Product Details