Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081119.1 | NP_001074588.1 | 2 | 3122 | |
NM_026868.1 | NP_081144.1 | 3 | 3229 | ||
GenBank mRNA | AK032495.1 | - | 1 | 1171 | |
AK040929.1 | - | 1 | 839 | ||
AK044407.1 | - | 2 | 2976 |