Gene Transcripts

Gene Symbol
Pgpep1l
Entrez Gene ID
78444
Gene Name
pyroglutamyl-peptidase I-like
Gene Aliases
C330024D12Rik
Chromosome Location
Chr. 7: 68236450 - 68264484 on Build GRCm38
UniGene ID
Mm.425468
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030101.1 NP_084377.1 4 619
GenBank mRNA AK021221.1 - 4 620

View Full Product Details