Gene Transcripts

Gene Symbol
Pik3r2
Entrez Gene ID
29741
Gene Name
phosphoinositide-3-kinase, regulatory subunit 2 (beta)
Gene Aliases
Chromosome Location
Chr. 16: 20416432 - 20424982 on Build Rnor_6.0
UniGene ID
Rn.22497
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022185.2 NP_071521.2 10 1544
XM_008771096.1 XP_008769318.1 9 2704
GenBank mRNA BC089805.1 - 10 1544
D64046.1 - 9 1104

View Full Product Details