Gene Transcripts
- Gene Symbol
- Sult1b1
- Entrez Gene ID
- 64305
- Gene Name
- sulfotransferase family, cytosolic, 1B, member 1
- Gene Aliases
- ST1B1
- Chromosome Location
- Chr. 14: 22142412 - 22155246 on Build Rnor_6.0
- UniGene ID
- Rn.1183
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022513.2 |
NP_071958.1
|
1, 2 |
243 |
|
XM_008770056.1 |
XP_008768278.1
|
1, 2 |
117 |
|
GenBank mRNA |
D89375.1 |
- |
1, 2 |
194 |
|
FQ209476.1 |
- |
1, 2 |
212 |
|
FQ209486.1 |
- |
1, 2 |
277 |
|
FQ209579.1 |
- |
1, 2 |
298 |
|
FQ210801.1 |
- |
1, 2 |
285 |
|
FQ212608.1 |
- |
1, 2 |
254 |
|
FQ218295.1 |
- |
1, 2 |
256 |
|
U38419.1 |
- |
1, 2 |
246 |
|
View Full Product Details