Gene Transcripts
- Gene Symbol
- Phkg1
- Entrez Gene ID
- 29353
- Gene Name
- phosphorylase kinase, gamma 1
- Gene Aliases
- PHKIN01, Phkg
- Chromosome Location
- Chr. 12: 30450431 - 30464314 on Build Rnor_6.0
- UniGene ID
- Rn.10399
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031573.1 |
NP_113761.1
|
3 |
188 |
|
GenBank mRNA |
X07320.1 |
- |
3 |
188 |
|
View Full Product Details