Gene Transcripts

Gene Symbol
Ybx1
Entrez Gene ID
500538
Gene Name
Y box binding protein 1
Gene Aliases
Byb1, Cbfa, Dbpb, Ef1a, Msy1, Nsep1, Yb1
Chromosome Location
Chr. 5: 138319753 - 138336721 on Build Rnor_6.0
UniGene ID
Rn.110976
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031563.3 NP_113751.3 3 368
XM_008764058.1 XP_008762280.1 3 408
XM_008764059.1 XP_008762281.1 3 608
GenBank mRNA AB046928.2 - 3 527
BC072486.1 - 3 368 7105037
BC098672.1 - 3 368 7124218
BC103637.1 - 3 362 7107271
D13309.1 - 3 338
M57299.1 - 3 344
M69138.1 - 3 504

View Full Product Details