Gene Transcripts

Gene Symbol
Trim3
Entrez Gene ID
83616
Gene Name
tripartite motif-containing 3
Gene Aliases
Berp, Rnf22
Chromosome Location
Chr. 1: 170441662 - 170464341 on Build Rnor_6.0
UniGene ID
Rn.203982
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031786.1 NP_113974.1 6 963
GenBank mRNA AF036255.1 - 6 963

View Full Product Details