Gene Transcripts

Gene Symbol
RNF11
Entrez Gene ID
26994
Gene Name
ring finger protein 11
Gene Aliases
CGI-123, SID1669
Chromosome Location
Chr. 1: 51236273 - 51273447 on Build GRCh38
UniGene ID
Hs.309641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014372.4 NP_055187.1 1, 2 615
GenBank mRNA AB024703.1 - 1, 2 258
AF151881.1 - 1, 2 258
AK293047.1 - 3, 4 444
AK313140.1 - 1, 2 377
BC020964.2 - 1, 2 210 3917224
BC047654.1 - 1, 2 583 5261966
DB265274.1 - 3, 4 444

View Full Product Details